9-(2-Biphenylyl)-10-bromoanthracene 98%

Supplier: Ambeed
Warning

Synonyms: 9-([1,1'-Biphenyl]-2-yl)-10-bromoanthracene

A326368-100MG A326368-1G A326368-250MG A326368-5G
77099-922EA 5 USD
77099-922 77099-924 77099-926 77099-928
9-(2-Biphenylyl)-10-bromoanthracene 98%
9-(2-Biphenylyl)-10-bromoanthracene

Precaución: for Research Use Only

Formula: C₂₆H₁₇Br
MW: 409.33 g/mol
MDL Number: MFCD28138083
CAS Number: 400607-16-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR