Methyl 4-amino-2,3-difluoro-5-nitrobenzoate 97%

Supplier: Ambeed
Warning

A326156-1G A326156-250MG A326156-25G A326156-5G
77099-914EA 24 USD
77099-914 77099-916 77099-918 77099-920
Methyl 4-amino-2,3-difluoro-5-nitrobenzoate 97%
Methyl 4-amino-2,3-difluoro-5-nitrobenzoate

Precaución: for Research Use Only

Formula: C₈H₆F₂N₂O₄
MDL Number: MFCD06201870
CAS Number: 284030-58-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR