(2,4,6-Trifluorophenyl)acetonitrile 98%

Supplier: Ambeed
Danger

A326142-1G A326142-250MG A326142-25G A326142-5G
77099-906EA 15 USD
77099-906 77099-908 77099-910 77099-912
(2,4,6-Trifluorophenyl)acetonitrile 98%
(2,4,6-Trifluorophenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₈H₄F₃N
MDL Number: MFCD00061221
CAS Number: 220227-80-5
UN: 3276
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR