3-Benzyloxybenzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: (3-(Benzyloxy)phenyl)methanol

A325444-100G A325444-1G A325444-25G A325444-5G
77099-898EA 249 USD
77099-898 77099-900 77099-902 77099-904
3-Benzyloxybenzyl alcohol 98%
3-Benzyloxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₁₄H₁₄O₂
MW: 214.26
Boiling Pt: 160
Melting Pt: 48
MDL Number: MFCD00004642
CAS Number: 1700-30-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR