6-Chloroquinoline-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A325290-250MG A325290-1G A325290-100MG
77099-896EA 194.15 USD
77099-896 77593-924 77685-930
6-Chloroquinoline-2-carboxylic acid 95%
6-Chloroquinoline-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₆ClNO₂
MW: 207.61 g/mol
MDL Number: MFCD09834123
CAS Number: 59394-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR