4-Phenoxypyridine 95%

Supplier: Ambeed
Warning

A323907-1G A323907-5G
77099-884EA 8.49 USD
77099-884 77099-886
4-Phenoxypyridine 95%
4-Phenoxypyridine

Precaución: for Research Use Only

Formula: C₁₁H₉NO
MW: 171.2000 g/mol
MDL Number: MFCD00235157
CAS Number: 4783-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR