(2-Bromo-5-fluorophenyl)hydrazine hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A321941-100MG
A321941-1G
A321941-250MG
A321941-5G
A321941-10G
77099-860EA
11
USD
77099-860
77099-862
77099-864
77593-912
77685-922
(2-Bromo-5-fluorophenyl)hydrazine hydrochloride 97%
(2-Bromo-5-fluorophenyl)hydrazine hydrochloride
Precaución: for Research Use Only
Formula:
C₆H₇BrClFN₂ |
MDL Number:
MFCD22571157 CAS Number: 60481-35-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...