6-Butoxynaphthalene-2-boronic acid 98%

Supplier: Ambeed
Warning

A319908-1G A319908-250MG A319908-5G
77099-848EA 81 USD
77099-848 77099-850 77099-852
6-Butoxynaphthalene-2-boronic acid 98%
6-Butoxynaphthalene-2-boronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₇BO₃
MW: 244.09 g/mol
MDL Number: MFCD16260721
CAS Number: 145369-28-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR