6-Bromo-1-isopropyl-1H-indazole-4-carboxylic acid 97%

Supplier: Ambeed
Warning

A319544-250MG A319544-100MG A319544-1G
77099-840EA 88.06 USD
77099-840 77685-915 77685-916
6-Bromo-1-isopropyl-1H-indazole-4-carboxylic acid 97%
6-Bromo-1-isopropyl-1H-indazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₁BrN₂O₂
MW: 283.12 g/mol
MDL Number: MFCD26407123
CAS Number: 1346702-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR