N-Methyl-1-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanamine 98%

Supplier: Ambeed
Warning

A319193-100MG A319193-1G A319193-250MG A319193-5G
77099-824EA 44.56 USD
77099-824 77099-826 77099-828 77099-830
N-Methyl-1-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanamine 98%
N-Methyl-1-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanamine

Precaución: for Research Use Only

Formula: C₁₄H₂₂BNO₂
MW: 247.14 g/mol
MDL Number: MFCD18733526
CAS Number: 1613259-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR