4-Chloro-1H-pyrazole 97%

Supplier: Ambeed
Warning

A317733-100G A317733-10G A317733-25G A317733-500G A317733-5G
77099-782EA 44 USD
77099-782 77099-784 77099-786 77099-788 77099-790
4-Chloro-1H-pyrazole 97%
4-Chloro-1H-pyrazole

Precaución: for Research Use Only

Formula: C₃H₃ClN₂
MDL Number: MFCD00159634
CAS Number: 15878-00-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR