(3-((tert-Butoxycarbonyl)(butyl)amino)phenyl)boronic acid 98%

Supplier: Ambeed
Warning

A315991-100MG A315991-1G A315991-250MG A315991-5G
77099-768EA 36.08 USD
77099-768 77099-770 77099-772 77099-774
(3-((tert-Butoxycarbonyl)(butyl)amino)phenyl)boronic acid 98%
(3-((tert-Butoxycarbonyl)(butyl)amino)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₅H₂₄BNO₄
MW: 293.16 g/mol
MDL Number: MFCD22571932
CAS Number: 925932-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR