2-Fluoro-1,3-dimethyl-4-nitrobenzene 95%

Supplier: Ambeed
Warning

A315070-1G A315070-250MG A315070-5G
77099-760EA 33 USD
77099-760 77099-762 77099-764
2-Fluoro-1,3-dimethyl-4-nitrobenzene 95%
2-Fluoro-1,3-dimethyl-4-nitrobenzene

Precaución: for Research Use Only

Formula: C₈H₈FNO₂
MDL Number: MFCD22683872
CAS Number: 1736-84-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR