2,4-Dichloro-6-fluoroquinoline 98%

Supplier: Ambeed
Warning

A313401-100MG A313401-1G A313401-250MG A313401-5G
77099-736EA 7 USD
77099-736 77099-738 77099-740 77593-870
2,4-Dichloro-6-fluoroquinoline 98%
2,4-Dichloro-6-fluoroquinoline

Precaución: for Research Use Only

Formula: C₉H₄Cl₂FN
MDL Number: MFCD09261383
CAS Number: 406204-74-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR