5-Ethynyl-2'-deoxycytidine 98%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-5-ethynyl-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one, 5'-Ethynyl-2'-deoxycytidine, EdC

A311059-100MG A311059-50MG
77099-714EA 118.83 USD
77099-714 77099-716
5-Ethynyl-2'-deoxycytidine 98%
5-Ethynyl-2'-deoxycytidine

Precaución: for Research Use Only

Formula: C₁₁H₁₃N₃O₄
MW: 251.24 g/mol
MDL Number: MFCD22417249
CAS Number: 69075-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR