2,4,5-Trifluorobenzyl bromide 97%

Supplier: Ambeed
Danger

Synonyms: 1-(Bromomethyl)-2,4,5-trifluorobenzene, α-Bromo-2,4,5-trifluorotoluene

A310038-1G A310038-25G A310038-5G
77099-694EA 6.37 USD
77099-694 77099-696 77099-698
2,4,5-Trifluorobenzyl bromide 97%
2,4,5-Trifluorobenzyl bromide

Precaución: for Research Use Only

Formula: C₇H₄BrF₃
MW: 225.01 g/mol
MDL Number: MFCD00061209
CAS Number: 157911-56-3
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR