3-(4-(Ethoxycarbonyl)-5-methyl-1H-pyrrol-3-yl)propanoic acid 97%

Supplier: Ambeed
Warning

A309268-1G A309268-250MG
77099-688EA 112.46 USD
77099-688 77099-690
3-(4-(Ethoxycarbonyl)-5-methyl-1H-pyrrol-3-yl)propanoic acid 97%
3-(4-(Ethoxycarbonyl)-5-methyl-1H-pyrrol-3-yl)propanoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₄
MW: 225.24 g/mol
MDL Number: MFCD09833175
CAS Number: 38664-16-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR