4-(3-Hydroxypropylcarbamoyl)phenylboronic acid pinacol ester 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-(3-Hydroxy-propyl)-4-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-benzamide, N-(3-Hydroxypropyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
A308494-1G
A308494-250MG
A308494-5G
77099-676EA
72.15
USD
77099-676
77099-678
77099-680
4-(3-Hydroxypropylcarbamoyl)phenylboronic acid pinacol ester 96%
4-(3-Hydroxypropylcarbamoyl)phenylboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₆H₂₄BNO₄ MW: 305.1800 g/mol |
MDL Number:
MFCD16294505 CAS Number: 1449134-30-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...