Fmoc-Sta-OH 95%

Supplier: Ambeed
Warning

Synonyms: (3S,4S)-4-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-hydroxy-6-methylheptanoic acid

A307157-1G A307157-250MG A307157-25G A307157-5G
77099-660EA 37.14 USD
77099-660 77099-662 77099-664 77099-666
Fmoc-Sta-OH 95%
Fmoc-Sta-OH

Precaución: for Research Use Only

Formula: C₂₃H₂₇NO₅
MW: 397.48 g/mol
MDL Number: MFCD00065675
CAS Number: 158257-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR