3-Bromo-4-chloroanisole 97%

Supplier: Ambeed
Warning

A306991-5G A306991-25G A306991-100G A306991-10G
77099-656EA 5.31 USD
77099-656 77099-658 77612-766 77901-810 77900-837
3-Bromo-4-chloroanisole 97%
3-Bromo-4-chloroanisole

Precaución: for Research Use Only

Formula: C₇H₆BrClO
MDL Number: MFCD00070739
CAS Number: 2732-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR