2-Amino-2-oxazoline hydrochloride 95%

Supplier: Ambeed
Danger

A304389-100MG A304389-1G A304389-250MG
77099-612EA 20 USD
77099-612 77099-614 77099-616
2-Amino-2-oxazoline hydrochloride 95%
2-Amino-2-oxazoline hydrochloride

Precaución: for Research Use Only

Formula: C₃H₆N₂O
MDL Number: MFCD00060178
CAS Number: 375855-07-5
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR