4-Fluoro-2-(trifluoromethyl)benzyl alcohol 97%

Supplier: Ambeed
Warning

Synonyms: (4-Fluoro-2-(trifluoromethyl)phenyl)methanol

A303561-1G A303561-250MG A303561-5G
77099-604EA 25 USD
77099-604 77099-606 77685-870
4-Fluoro-2-(trifluoromethyl)benzyl alcohol 97%
4-Fluoro-2-(trifluoromethyl)benzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₆F₄O
MDL Number: MFCD00061170
CAS Number: 220227-29-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR