Methyl (S)-(-)-lactate 98%

Supplier: Ambeed
Danger

Synonyms: (S)-Methyl 2-hydroxypropanoate, Methyl (S)-(-)-2-hydroxypropionate, Sodium carboxymethyl cellulose

A303264-25G A303264-100G
77099-602EA 8.49 USD
77099-602 77612-746
Methyl (S)-(-)-lactate 98%
Methyl (S)-(-)-lactate

Precaución: for Research Use Only

Formula: C₄H₈O₃
MW: 104.11 g/mol
MDL Number: MFCD00064265
CAS Number: 27871-49-4
UN: 3272
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR