2,4,6-Trifluorobenzylamine 95%

Supplier: Ambeed
Warning

A303259-100G A303259-10G A303259-1G A303259-250MG A303259-25G A303259-5G
77099-590EA 291 USD
77099-590 77099-592 77099-594 77099-596 77099-598 77099-600
2,4,6-Trifluorobenzylamine 95%
2,4,6-Trifluorobenzylamine

Precaución: for Research Use Only

Formula: C₇H₆F₃N
MDL Number: MFCD00236317
CAS Number: 214759-21-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR