2-Bromo-6-iodoaniline 97%

Supplier: Ambeed
Warning

A303076-100MG A303076-1G A303076-5G
77099-580EA 21 USD
77099-580 77612-744 77685-866
2-Bromo-6-iodoaniline 97%
2-Bromo-6-iodoaniline

Precaución: for Research Use Only

Formula: C₆H₅BrIN
MDL Number: MFCD18391615
CAS Number: 84483-27-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR