Ethyl 2,2-Difluoro-2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetate 98%

Supplier: Ambeed
Warning

A302744-100MG A302744-1G A302744-250MG
77099-574EA 37.14 USD
77099-574 77099-576 77099-578
Ethyl 2,2-Difluoro-2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetate 98%
Ethyl 2,2-Difluoro-2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetate

Precaución: for Research Use Only

Formula: C₁₆H₂₁BF₂O₄
MW: 326.14 g/mol
MDL Number: MFCD20278276
CAS Number: 1683545-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR