1-Iodo-2-naphthol 97%

Supplier: Ambeed
Warning

A302526-10G A302526-1G A302526-250MG A302526-5G A302526-25G
77099-566EA 257 USD
77099-566 77099-568 77099-570 77099-572 77901-182
1-Iodo-2-naphthol 97%
1-Iodo-2-naphthol

Precaución: for Research Use Only

Formula: C₁₀H₇IO
MDL Number: MFCD00046371
CAS Number: 2033-42-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR