9-Phenyl-3,6-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole 97%

Supplier: Ambeed
Warning

A302502-1G A302502-250MG A302502-5G
77099-560EA 30.77 USD
77099-560 77099-562 77099-564
9-Phenyl-3,6-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole 97%
9-Phenyl-3,6-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole

Precaución: for Research Use Only

Formula: C₃₀H₃₅B₂NO₄
MW: 495.22 g/mol
MDL Number: MFCD28015750
CAS Number: 618442-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR