3-Methyl-4-nitrobenzaldehyde 97%
Supplier: Ambeed
|
Warning
|
A302251-100MG
A302251-10G
A302251-1G
A302251-250MG
77099-552EA
10.61
USD
77099-552
77099-554
77099-556
77099-558
3-Methyl-4-nitrobenzaldehyde 97%
3-Methyl-4-nitrobenzaldehyde
Precaución: for Research Use Only
Formula:
C8H7NO3 MW: 165.1500 g/mol |
MDL Number:
MFCD08445621 CAS Number: 18515-67-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...