6-Bromo-5-fluoro-1,2,3,4-tetrahydroisoquinoline 95%
Supplier: Ambeed
|
Warning
|
A301530-100MG
A301530-1G
A301530-250MG
77099-540EA
151.71
USD
77099-540
77099-542
77099-544
6-Bromo-5-fluoro-1,2,3,4-tetrahydroisoquinoline 95%
6-Bromo-5-fluoro-1,2,3,4-tetrahydroisoquinoline
Precaución: for Research Use Only
Formula:
C₉H₉BrFN MW: 230.07 g/mol |
MDL Number:
MFCD28991843 CAS Number: 1781658-50-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...