6-Azaspiro[3.4]octan-5-one 98%

Supplier: Ambeed
Warning

A301390-100MG A301390-1G A301390-250MG
77099-530EA 89 USD
77099-530 77099-532 77099-534
6-Azaspiro[3.4]octan-5-one 98%
6-Azaspiro[3.4]octan-5-one

Precaución: for Research Use Only

Formula: C₇H₁₁NO
MDL Number: MFCD19219361
CAS Number: 1193-30-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR