Methyl-2-(chloromethyl)thiazole-4-carboxylate 97%

Supplier: Ambeed
Danger

Synonyms: Methyl 2-(chloromethyl)thiazole-4-carboxylate

A300836-1G A300836-250MG A300836-5G
77099-522EA 70.02 USD
77099-522 77099-524 77099-526
Methyl-2-(chloromethyl)thiazole-4-carboxylate 97%
Methyl-2-(chloromethyl)thiazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₆H₆ClNO₂S
MW: 191.6400 g/mol
MDL Number: MFCD08669440
CAS Number: 321371-29-3
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR