Sodium 3-bromobenzenesulfinate 90%

Supplier: Ambeed
Warning

A300520-100MG A300520-1G A300520-250MG
77099-516EA 39 USD
77099-516 77099-518 77099-520
Sodium 3-bromobenzenesulfinate 90%
Sodium 3-bromobenzenesulfinate

Precaución: for Research Use Only

Formula: C₆H₄BrNaO₂S
MDL Number: MFCD11618015
CAS Number: 89520-70-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR