5-Bromo-2-chloro-4-fluorophenylacetonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 2-(5-Bromo-2-chloro-4-fluorophenyl)acetonitrile, 5-Bromo-2-chloro-4-fluorobenzyl cyanide

A300441-100MG A300441-1G A300441-250MG A300441-25G
77099-508EA 5.31 USD
77099-508 77099-510 77099-512 77099-514
5-Bromo-2-chloro-4-fluorophenylacetonitrile 97%
5-Bromo-2-chloro-4-fluorophenylacetonitrile

Precaución: for Research Use Only

Formula: C₈H₄BrClFN
MW: 248.479 g/mol
MDL Number: MFCD23166759
CAS Number: 1426290-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR