1-(Methylsulphonyl)piperazine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Methanesulphonylpiperazine
A300303-100G
A300303-10G
A300303-1G
A300303-25G
A300303-500G
A300303-5G
77099-488EA
213
USD
77099-488
77099-490
77099-492
77099-494
77099-496
77099-498
1-(Methylsulphonyl)piperazine 97%
1-(Methylsulphonyl)piperazine
Precaución: for Research Use Only
Formula:
C₅H₁₂N₂O₂S |
MDL Number:
MFCD02153225 CAS Number: 55276-43-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...