4-Biphenylyl trifluoromethanesulfonate 95%

Supplier: Ambeed
Warning

Synonyms: 4-Phenylphenyl triflate, 4-Biphenylyl triflate, Trifluoromethanesulphonic acid 4-Biphenylyl ester

A297991-1G A297991-250MG A297991-5G
77099-460EA 25 USD
77099-460 77099-462 77099-464
4-Biphenylyl trifluoromethanesulfonate 95%
4-Biphenylyl trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₁₃H₉F₃O₃S
MW: 302.27 g/mol
MDL Number: MFCD18379753
CAS Number: 17763-78-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR