Potassium (3,4-difluorophenyl)trifluoroborate 95%

Supplier: Ambeed
Warning

A297629-1G A297629-250MG A297629-5G
77099-450EA 72 USD
77099-450 77099-452 77099-454
Potassium (3,4-difluorophenyl)trifluoroborate 95%
Potassium (3,4-difluorophenyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₆H₃BF₅K
MW: 219.99 g/mol
MDL Number: MFCD09993188
CAS Number: 1033592-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR