2-Amino-5-bromo-4-fluorophenol 97%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-5-fluoro-2-hydroxyaniline

A296942-1G A296942-250MG A296942-10G A296942-5G
77099-440EA 127.31 USD
77099-440 77099-442 77685-844 77685-845
2-Amino-5-bromo-4-fluorophenol 97%
2-Amino-5-bromo-4-fluorophenol

Precaución: for Research Use Only

Formula: C₆H₅BrFNO
MW: 206.012 g/mol
MDL Number: MFCD24640741
CAS Number: 1037364-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR