2-(4-Chloro-2,3-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-2,3-difluorophenylboronic acid pinacol ester

A296941-1G A296941-250MG A296941-25G A296941-5G
77099-432EA 158.08 USD
77099-432 77099-434 77099-436 77099-438
2-(4-Chloro-2,3-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%
2-(4-Chloro-2,3-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₂H₁₄BClF₂O₂
MW: 274.50 g/mol
MDL Number: MFCD16659094
CAS Number: 1116681-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR