Cimetidine 98%

Supplier: Ambeed
Danger

A295450-100G A295450-1G A295450-25G A295450-500G A295450-5G A295450-1MG
77099-414EA 101 USD
77099-414 77099-416 77099-418 77099-420 77099-422 77099-424 77685-839
Cimetidine 98%
Cimetidine

Precaución: for Research Use Only

Formula: C₁₀H₁₆N₆S
MDL Number: MFCD00133296
CAS Number: 51481-61-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR