3-Bromophenetole 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromophenyl ethyl ether, 1-Bromo-3-ethoxybenzene

A295336-1G A295336-25G A295336-5G
77099-408EA 10 USD
77099-408 77099-410 77099-412
3-Bromophenetole 98%
3-Bromophenetole

Precaución: for Research Use Only

Formula: C₈H₉BrO
MW: 201.06 g/mol
MDL Number: MFCD00274287
CAS Number: 2655-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR