L(+)-Cysteine hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: H-Cys-OH.HCl, (R)-2-Amino-3-mercaptopropanoic acid hydrochloride, L-Cysteine hydrochloride, (R)-(+)-Cysteine hydrochloride

A295232-1000G A295232-100G A295232-25G A295232-500G A295232-50MG
77099-398EA 81 USD
77099-398 77099-400 77099-402 77099-404 77099-406
L(+)-Cysteine hydrochloride 95%
L(+)-Cysteine hydrochloride

Precaución: for Research Use Only

Formula: HSCH₂CH(NH₂)COOH·HCl
MW: 157.62 g/mol
Melting Pt: 175
MDL Number: MFCD00064553
CAS Number: 52-89-1
Merck Index: 13,02810

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR