Boc-Dab(Boc)-OH·DCHA 95%

Supplier: Ambeed
Warning

Synonyms: Boc-Dab(Boc)-OH·DCHA, Boc-Dab(Boc)-OH.DCHA, (R)-2,3-Di-(Boc-amino)propionic acid dicyclohexylammonium salt

A294433-1G A294433-5G
77099-390EA 35 USD
77099-390 77099-392 77099-394
Boc-Dab(Boc)-OH·DCHA 95%
Boc-Dab(Boc)-OH·DCHA

Precaución: for Research Use Only

Formula: C₂₆H₄₉N₃O₆
MW: 499.693 g/mol
MDL Number: MFCD00238312
CAS Number: 201472-66-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR