3,4-Dibutoxy-3-cyclobutene-1,2-dione 98%

Supplier: Ambeed
Warning

Synonyms: Squaric acid dibutylester, Dibutyl squarate, 3,4-Dibutoxycyclobut-3-ene-1,2-dione, 1,2-Dibutoxy-1-cyclobutene-3,4-dione

A294391-10G A294391-1G A294391-25G A294391-5G A294391-100G
77099-382EA 37 USD
77099-382 77099-384 77099-386 77099-388 77900-688
3,4-Dibutoxy-3-cyclobutene-1,2-dione 98%
3,4-Dibutoxy-3-cyclobutene-1,2-dione

Precaución: for Research Use Only

Formula: C₁₂H₁₈O₄
MW: 226.27 g/mol
Boiling Pt: 121
MDL Number: MFCD00037150
CAS Number: 2892-62-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR