1-(3-Chlorophenyl)-1H-pyrazole-4-boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 1-(3-Chlorophenyl)pyrazole-4-boronic acid

A290989-100MG A290989-1G A290989-250MG
77099-324EA 36.08 USD
77099-324 77099-326 77099-328
1-(3-Chlorophenyl)-1H-pyrazole-4-boronic acid 98%
1-(3-Chlorophenyl)-1H-pyrazole-4-boronic acid

Precaución: for Research Use Only

Formula: C₉H₈BClN₂O₂
MW: 222.4400 g/mol
MDL Number: MFCD09878353
CAS Number: 1072945-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR