4-Butylthiophenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-(Butylthio)phenyl)boronic acid

A289029-1G A289029-5G
77099-294EA 18.04 USD
77099-294 77099-296
4-Butylthiophenylboronic acid 98%
4-Butylthiophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₅BO₂S
MW: 210.1 g/mol
MDL Number: MFCD06801729
CAS Number: 151588-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR