2-(Methylsulfonyl)-2,4,5,6-tetrahydropyrrolo[3,4-c]pyrazole 95%

Supplier: Ambeed
Warning

A287627-100MG A287627-250MG
77099-284EA 100.79 USD
77099-284 77099-286
2-(Methylsulfonyl)-2,4,5,6-tetrahydropyrrolo[3,4-c]pyrazole 95%
2-(Methylsulfonyl)-2,4,5,6-tetrahydropyrrolo[3,4-c]pyrazole

Precaución: for Research Use Only

Formula: C₆H₉N₃O₂S
MW: 187.21 g/mol
MDL Number: MFCD20687608
CAS Number: 1226781-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR