2-(3,5-Dibromophenyl)-4,6-diphenyl-1,3,5-triazine 97%

Supplier: Ambeed
Warning

A287569-100MG A287569-1G A287569-250MG A287569-5G
77099-276EA 31.83 USD
77099-276 77099-278 77099-280 77099-282
2-(3,5-Dibromophenyl)-4,6-diphenyl-1,3,5-triazine 97%
2-(3,5-Dibromophenyl)-4,6-diphenyl-1,3,5-triazine

Precaución: for Research Use Only

Formula: C₂₁H₁₃Br₂N₃
MW: 467.16 g/mol
MDL Number: MFCD25562933
CAS Number: 1073062-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR