1-Butyl-3-methylpyridinium chloride 98%

Supplier: Ambeed
Warning

Synonyms: 1-Butyl-3-methylpyridinium Chloride

A287216-100G A287216-1G A287216-25G A287216-5G
77099-266EA 354.35 USD
77099-266 77099-268 77099-270 77099-272
1-Butyl-3-methylpyridinium chloride 98%
1-Butyl-3-methylpyridinium chloride

Precaución: for Research Use Only

Formula: C₁₀H₁₆ClN
MW: 185.7 g/mol
MDL Number: MFCD08458942
CAS Number: 125652-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR