2,6-Dibromo-4-fluorobenzaldehyde 95%

Supplier: Ambeed
Danger

A286805-100MG A286805-1G A286805-250MG A286805-25G A286805-5G
77099-256EA 21.22 USD
77099-256 77099-258 77099-260 77099-262 77099-264
2,6-Dibromo-4-fluorobenzaldehyde 95%
2,6-Dibromo-4-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃Br₂FO
MW: 281.90 g/mol
MDL Number: MFCD13186760
CAS Number: 938467-02-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR